Wheat Germ Agglutinin (WGA)
Invitrogen™

Wheat Germ Agglutinin (WGA)

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) and Alexa Fluor, Alexa Fluor Plus, and other dyes. Fluorescent WGA conjugates bind to carbohydrates and are used for various cell biology applications such as plasma membrane labeling and cell painting assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugate
W21405Texas Red-X
W11263Alexa Fluor 350
W7024Alexa Fluor 350, Oregon Green 488, Tetramethylrhodamine, Texas Red
W11261Alexa Fluor 488
W32464Alexa Fluor 555
W11262Alexa Fluor 594
W21404Alexa Fluor 633
W32466Alexa Fluor 647
W32465Alexa Fluor 680
W56132Alexa Fluor Plus 405
W56133Alexa Fluor Plus 568
W56134Alexa Fluor Plus 770
W834Fluorescein
W6748Oregon Green 488
Q12021MPQdot 655
W849Tetramethylrhodamine
Catalog number W21405
Price (BRL)
1.783,43
1 mg
Conjugate:
Texas Red-X
Price (BRL)
1.783,43
1 mg

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates. These lectins can bind to carbohydrates and are available conjugated to Alexa Fluor™, Alexa Fluor™ Plus, and other fluorescent dyes. Fluorescent wheat germ agglutinin conjugates are valuable tools in molecular and cell biology research, enabling researchers to label the plasma membrane in fluorescence imaging and cell painting assays, and study and analyze glycosylation patterns and glycan-mediated processes in cells and tissues.

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) with Alexa Fluor, Alexa Fluor Plus, and other dyes. WGA is a cell impermeant stain that selectively binds to N‐acetylglucosamine and N‐acetylneuraminic acid (sialic acid) residues, which are often found on cell membranes. Fluorescent WGA conjugates provide selective labeling of the plasma membrane with minimal background in many cell types that is retained after formaldehyde fixation and permeabilization with Triton X-100.

These fluorescent lectin conjugates can also be used to label fixed cells; however, to avoid labeling intracellular components, formaldehyde-fixed cells should not be permeabilized before labeling. Fluorescent WGA conjugates are used as plasma membrane stains along with other cellular markers in cell painting assays to provide a phenotypic readout of cell health or cytotoxicity. The Wheat Germ Agglutinin, Alexa Fluor 555 Conjugate is included in the Image-iT Cell Painting Kit (Cat. Nos. I65000 and I65500).

WGA conjugates are also used as retrograde tracers for neuronal tracing experiments and have been shown to cross synapses. These fluorescent lectins are applicable in microbiology studies to label yeast bud scars, the cell membrane of gram-positive but not gram-negative bacteria, and chitin in fungal cell walls. In solution, WGA exists as a heterodimer with a molecular weight of approximately 38,000 Daltons and is normally cationic under physiological conditions. Our WGA conjugates have been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), flow cytometry (FC), and a wide range of chemical, biochemical and immunological assays.

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates with options covering the entire wavelength range. The Wheat Germ Agglutinin Sampler Kit (Cat. No. W7024) includes introductory samples of four fluorescent WGAs: Alexa Fluor 350, Oregon Green 488, tetramethylrhodamine, and Texas Red-X conjugates. The red-fluorescent Alexa Fluor 594 wheat germ agglutinin (WGA) conjugate is also included in the Image-iT LIVE Plasma Membrane and Nuclear Labeling Kit (Cat. No. I34406). The Texas Red-X WGA conjugate can be purchased in the ViaGram Red+ Bacterial Gram Stain and Viability Kit (Cat. No. V7023) to differentiate gram-positive and gram-negative bacteria.

Wheat germ agglutinin (WGA) conjugates fluorescence excitation/emission
W11261 WGA, Alexa Fluor 488 conjugate: 495 nm/519 nm
W11263 WGA, Alexa Fluor 350 conjugate: 346 nm/442 nm
W32464 WGA, Alexa Fluor 555 conjugate: 555 nm/580 nm
W11262 WGA, Alexa Fluor 594 conjugate: 590 nm/617 nm
W21404 WGA, Alexa Fluor 633 conjugate: 632 nm/647 nm
W32466 WGA, Alexa Fluor 647 conjugate: 650 nm/665 nm
W32465 WGA, Alexa Fluor 680 conjugate: 679 nm/702 nm
W56132 WGA, Alexa Fluor Plus 405 conjugate: 408 nm/450 nm
W56133 WGA, Alexa Fluor Plus 568 conjugate: 562 nm/583 nm
W56134 WGA, Alexa Fluor Plus 770 conjugate: 770 nm/797 nm
W834 WGA, fluorescein conjugate: 494 nm/518 nm
W6748 WGA, Oregon Green 488 conjugate: 496 nm/524 nm
W849 WGA, tetramethylrhodamine conjugate: 555 nm/580 nm
W21405 WGA, Texas Red-X conjugate: 595 nm/615 nm

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Common NameWheat Germ Agglutinin, Texas Red™-X Conjugate
Expression SystemWheat germ
Ligand TypeN-acetylglucosamine and N-acetylneuraminic acid (sialic acid) residues
Product LineTexas Red
ProteinFluorescent lectins
Protein FamilyLectins
Protein FormHeterodimer
Protein SubtypeAgglutinin
Protein TagNone
Purity or Quality GradeSee Certificate of Analysis
Quantity1 mg
Shipping ConditionRoom Temperature
SourceWheat germ
ConjugateTexas Red-X
For Use With (Application)Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Cell Painting
FormLyophilized
RecombinantNative
SpeciesWheat
Unit Size1 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

My cells are very sensitive and need to be kept in media as much as possible. Is it possible to label the plasma membrane with fluorescent wheat germ agglutinin (WGA) in media instead of buffer?

Yes. Although labeling in buffer (such as Hank's Buffered Saline Solution) is slightly better for brightness and lower non-cell background, media can be used. Do a concentration range to dertermine optimal conditions, since the WGA may potentially bind media components to some extent, slightly decreasing your specific labeling intensity.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use CellMask Plasma membrane stains or Alexa Fluor dye labeled wheat germ agglutinin to label the plasma membrane of my paraffin sections?

No. For paraffin sections, there are few options due to the delipidation of the membranes by solvents used in the deparaffinization steps. The only good option is to use an antibody against a plasma membrane protein.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to label Golgi bodies in my fixed cells. Is a dye-conjugated lectin like wheat germ agglutinin or concanavalin A a good choice?

No. Those lectins have been shown to label Golgi in fixed and permeabilized cultured cells, but the selectivity is cell type dependent. Usually you wind up with other structures labeling as well, such as endoplasmic reticulum. The only guaranteed way to specifically label Golgi in already fixed cells is to use an antibody for a Golgi-specific antigen.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to label the plasma membrane of my cells, but there are several dyes to choose from. Which one should I use?

For live-cell imaging, the CellVue and CellMask Plasma Membrane Stains are the most uniform and the slowest to be endocytosed. However, they are not the best choice if you wish to fix and permeabilize your cells, such as for antibody labeling. Wheat germ agglutinin (WGA) conjugates are also able to label live cells, or can label already formaldehyde-fixed cells. They can survive subsequent permeabilization with detergents, such as Triton X-100. If cells are already permeabilized, WGA will label internal structures as well. Thus, only an antibody against a plasma membrane protein can be used if cells are already permeabilized. Lipophilic cyanine dyes, such as DiI, will label all cell membranes in live cells, not just plasma membranes, if left on live cells for extended periods. Following page will help you choose (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-structure/plasma-membrane.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (33)

Citations & References
Abstract
Indirect immunofluorescence localization of ponticulin in motile cells.
Authors:Wuestehube LJ, Chia CP, Luna EJ
Journal:Cell Motil Cytoskeleton
PubMed ID:2673549
'Ponticulin is the major actin-binding integral glycoprotein in plasma membranes isolated from log-phase Dictyostelium discoideum amebae. As such, this protein appears to be an important link between the plasma membrane and actin filaments (Wuestehube and Luna: Journal of Cell Biology 105:1741-1751, 1987). In this study, indirect immunofluorescence microscopy was used ... More
A functional YNKI motif in the short cytoplasmic tail of varicella-zoster virus glycoprotein gH mediates clathrin-dependent and antibody-independent endocytosis.
Authors:Pasieka TJ, Maresova L, Grose C,
Journal:J Virol
PubMed ID:12634377
'The trafficking of varicella-zoster virus (VZV) gH was investigated under both infection and transfection conditions. In initial endocytosis assays performed in infected cells, the three glycoproteins gE, gI, and gB served as positive controls for internalization from the plasma membrane. Subsequently, we discovered that gH in VZV-infected cells was also ... More
Rotavirus NSP4 induces a novel vesicular compartment regulated by calcium and associated with viroplasms.
Authors:Berkova Z, Crawford SE, Trugnan G, Yoshimori T, Morris AP, Estes MK
Journal:J Virol
PubMed ID:16731945
'Rotavirus is a major cause of infantile viral gastroenteritis. Rotavirus nonstructural protein 4 (NSP4) has pleiotropic properties and functions in viral morphogenesis as well as pathogenesis. Recent reports show that the inhibition of NSP4 expression by small interfering RNAs leads to alteration of the production and distribution of other viral ... More
Isolation of bright aggregate fluctuations in a multipopulation image correlation spectroscopy system using intensity subtraction.
Authors:Rocheleau JV, Wiseman PW, Petersen NO
Journal:Biophys J
PubMed ID:12770905
'Image correlation spectroscopy allows sensitive measurement of the spatial distribution and aggregation state of fluorescent membrane macro molecules. When studying a single population system (i.e., aggregates of similar brightness), an accurate measure can be made of the aggregate number per observation area, but this measurement becomes much more complex in ... More
Microstructural differences between single-species and dual-species biofilms of Streptococcus mutans and Veillonella parvula, before and after exposure to chlorhexidine.
Authors:Kara D, Luppens SB, van Marle J, Ozok R, ten Cate JM,
Journal:FEMS Microbiol Lett
PubMed ID:17403046
'Dual-species biofilms of Streptococcus mutans and Veillonella parvula are less susceptible to antimicrobials than single-species biofilms of the same microorganisms. The microstructure of single and dual-species biofilms of S. mutans and/or V. parvula was visualized to find out whether the spatial arrangement of bacteria in biofilms is related to survival ... More